Smoothened agonist ≥95%

Supplier: Enzo Life Sciences

Synonyms: SAG3-Chloro-N-[trans-4-(methylamino)cyclohexyl]-N-[3-(4-pyridinyl)benzyl]-1-benzothiophene-2-carboxamide

270-426-M001
89147-870EA 299.16 USD
89147-870
Smoothened agonist ≥95%
Smoothened agonist

Smoothened ligand


A potent cell-permeable benzothiophene compound that modulates the coupling of Smo with its downstream effector by interacting with the Smo heptahelical domain (KD = 59 nM). Shown to induce Hedgehog pathway activation (EC50= ~3 nM in NIH 3T3-derived Shh-LIGHT2 cells) and counteracts Cyclopamine (BML-GR334) inhibition of Smo. Reported to act as an activator at low concentrations and as an inhibitor at very high concentrations.

Formula: C₂₈H₂₈ClN₃OS
MW: 490.062 g/mol
MDL Number: MFCD22581513
CAS Number: 912545-86-9
https://us.vwr.com/stibo/web/std.lang.all/75/62/45347562.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR