Tannic acid ≥95%

Supplier: Enzo Life Sciences

Synonyms: TanninGallotannin

270-418-G001
89147-850EA 51.84 USD
89147-850
Tannic acid ≥95%
Tannic acid

Inhibitor of PARG and eNOS


Inhibitor of poly(ADP-ribose) glycohydrolase (PARG). Cytoprotective in oxidatively stressed cells. Inhibitor of endothelial nitric oxide synthase (eNOS; NOS III) and weak inhibitor of inducible (iNOS; NOS II) and neuronal nitric oxide synthase (nNOS; NOS I). Induces cyclooxygenase-2 (COX-2) expression. Free radical scavenger.


Occurs in the bark and fruit of many plants, notably in the bark of the oak species, in sumac and myrobalan.

Formula: C₇₆H₅₂O₄₆
MW: 1701.2 g/mol
MDL Number: MFCD00066397
CAS Number: 1401-55-4
https://us.vwr.com/stibo/web/std.lang.all/75/57/45347557.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR