TIQ-A ≥96% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: Thieno[2,3-c]isoquinolin-5-one

270-365-M005 270-365-M001
89147-716EA 230 USD InStock
89147-716 89147-714
TIQ-A ≥96% (by HPLC)
TIQ-A

Potent poly(ADP-ribose) polymerase-1 (PARP-1) inhibitor (IC50=450nM). Neuroprotectant.


  • PARP-1 inhibitor


Neuroprotectant.


Soluble in DMSO.

Formula: C11H7NOS
MW: 201,2
Storage Temperature: Refrigerator
CAS Number: 420849-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR