3-O-Methyl-N-acetyl-D-glucosamine ≥98% (by 1H-NMR)
Synonyms:
2-Acetamido-2-deoxy-3-O-methyl-D-glucopyranose
, 3-OMe-GlcNAc
270-343-M005
89147-656EA
352.47
USD
89147-656
3-O-Methyl-N-acetyl-D-glucosamine ≥98% (by 1H-NMR)
3-O-Methyl-N-acetyl-D-glucosamine
N-Acetylglucosamine kinase inhibitor
Inhibitor of N-acetylglucosamine kinase (Ki=17µM) and N-acetylmannosamine kinase (Ki=80µM). It has been found that 3-OMe-GlcNAc potently inhibits glucose phosphorylation by N-acetylglucosamine kinase whereas glucokinase is not at all affected by this hexosamine.
Formula:
C₉H₁₇NO₆ MW: 235,2 g/mol |
CAS Number:
94825-74-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...