Raf1 Kinase Inhibitor I ≥97% (by HPLC)

Synonyms: 5-Iodo-3-[(3,5-dibromo-4-hydroxyphenyl)methylene]-2-indolinone3-(3,5-dibromo-4-hydroxybenzylidene)-5-iodoindolin-2-one

270-324-M001
89147-634EA 331.2 USD
89147-634
Raf1 Kinase Inhibitor I ≥97% (by HPLC)
Raf1 Kinase Inhibitor I

Potent Raf1 kinase inhibitor (IC50=9nM). Shows ≥100-fold selectivity for Raf kinase versus CDK1, CDK2, c-Src, ERK2, MEK, p38, Tie2, VEGFR2, and c-Fms.


Off-white to orange powder.


Soluble in DMSO (200mg/ml).

Formula: C₁₅H₈Br₂INO₂
MW: 520.9 g/mol
Storage Temperature: Freezer
CAS Number: 220904-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR