ASSA270-323
270-323-M001
89147-632EA 182.57 USD
89147-632
ASSA270-323
Potent, cell permeable, reversible, and selective inhibitor of EGF receptor (EGFR) tyrosine kinase activity (IC50=450pM).
  • EGFR inhibitor
Off white to yellow powder.
Soluble in DMSO (200 mg/ml).