PD 168,393 ≥95% (by HPLC)
Synonyms:
N-[4-[(3-bromophenyl)amino]-6-quinazolinyl]-2-propenamide
, 4-[(3-Bromophenyl)amino]-6-acrylamidoquinazoline
, N-(4-((3-Bromophenyl)amino)quinazolin-6-yl)acrylamide
, PD 168,393
270-322-M001
89147-630EA
317.93
USD
89147-630
PD 168,393 ≥95% (by HPLC)
PD-168393
Potent, cell permeable, irreversible, and selective inhibitor of EGF receptor (EGFR) tyrosine kinase activity (IC50=700pM). Antitumor agent in vivo.
- EGFR inhibitor
Yellow to orange solid.
Soluble in DMSO (200 mg/ml).
Formula:
C₁₇H₁₃BrN₄O MW: 369.2 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD02179207 CAS Number: 194423-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...