Formononetin ≥97% (by HPLC, TLC), high purity
Synonyms:
7-Hydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one
, 7-Hydroxy-3-(4-methoxyphenyl)chromone
, 7-Hydroxy-4'-methoxyisoflavone
270-312-M005
89147-620EA
240.27
USD
89147-620
Formononetin ≥97% (by HPLC, TLC), high purity
Formononetin
IL-4 activator
Interacts with human estrogen receptors with low potency. Enhances IL-4 production in a dose-dependent manner. Inhibits lecithin peroxidation induced by hydroxyl radicals. Selective inhibitor of the γ-isoform of alcohol dehydrogenase. Antioxidant.
Formula:
C₁₆H₁₂O₄ MW: 268.27 g/mol |
MDL Number:
MFCD00016948 CAS Number: 485-72-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...