6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic cid ≥96.5% (by titrimetric analysis)
Supplier: Enzo Life Sciences
Synonyms:
Trolox
, 6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic acid
270-267-M100
89147-528EA
50.86
USD
89147-528
6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic cid ≥96.5% (by titrimetric analysis)
6-Hydroxy-2,5,7,8-tetramethylchroman-2-carboxylic cid
Cell permeable derivative of vitamin E. Antioxidant. Inhibits peroxynitrite mediated tyrosine nitration and mitochondrial proton permeability. Prevents lipid peroxidation, cytotoxicity and apoptosis induced by cytochrome P450 2E1 in HEP G2 cells.
- Cell permeable vitamin E derivative
White to off-white, pale yellow or pale green powder.
Soluble in 100% ethanol (25mg/ml) or methanol.
Formula:
C₁₄H₁₈O₄ MW: 250.29 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00006846 CAS Number: 53188-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...