AM 281 ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Danger

270-240-M001 270-240-M025 270-240-M005
89147-472EA 369.77 USD
89147-472 89147-470 89147-468
AM 281 ≥98% (by HPLC)
AM 281

CB1 receptor ligand.


Analog of the cannabinoid receptor (CB) antagonist SR 141716A. Potent CB1 receptor antagonist/inverse agonist (Ki = 14 nM).


Soluble in methanol (1 mg/mL) or DMSO (2 mg/mL at RT, 50 mg/mL in hot DMSO, after cooling this solution is stable over several days).


Synonym: 1-(2,4-Dichlorophenyl)-5-(4-iodophenyl)-4-methyl-N-4-morpholinyl-1H-pyrazole-3-carboxamide

Formula: C₂₁H₁₉Cl₂IN₄O₂
MW: 557.2 g/mol
Storage Temperature: Refrigerator
CAS Number: 202463-68-1
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/74/95/45347495.jpg

Order Now

Specification Test Results

Purity ≥98% (HPLC)
Identity Determined by ¹H-NMR

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR