ASSA270-234
270-234-M005
89147-462EA 94.51 USD
89147-462
ASSA270-234
Calpain inhibitor
Cell permeable, selective, and non-peptide calpain inhibitor directed towards the calcium binding sites of calpain. Exhibits high specificity for calpains relative to other proteases such as cathepsin B and cathepsin L. Prevents dexamethasone-induced apoptosis in thymocytes. Calpain I (Ki=210nM), Calpain II (Ki=370nM)