Milrinone ≥97% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: 6-Methyl-5-(4-pyridyl)-2-pyridone-3-carbonitrile3-Cyano-6-methyl-5-(4-pyridyl)-2-pyridone2-Methyl-6-oxo-1,6-dihydro-[3,4'-bipyridine]-5-carbonitrile

270-083-M005
89147-182EA 143.84 USD
89147-182
Milrinone ≥97% (by TLC)
Milrinone

PDE3 inhibitor


Selective inhibitor of phosphodiesterase 3 (PDE3, IC50=0.3µM).

Formula: C₁₂H₉N₃O
MW: 211.22 g/mol
MDL Number: MFCD00133539
CAS Number: 78415-72-2
https://us.vwr.com/stibo/web/std.lang.all/74/73/45347473.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR