HA-100 dihydrochloride ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
5-(Piperazin-1-ylsulphonyl)isoquinoline
, 5-(Piperazin-1-ylsulfonyl)isoquinoline
270-069-M005
89147-168EA
116.82
USD
89147-168
HA-100 dihydrochloride ≥98% (by HPLC)
HA-100 dihydrochloride
Protein kinase inhibitor
Inhibitor of myosin light chain kinase, cAMP-dependent protein kinase (PKA) and protein kinase C (PKC).
Formula:
C₁₃H₁₅N₃O₂S MW: 277.3421 g/mol |
MDL Number:
MFCD00209852 CAS Number: 84468-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...