Tyrphostin AG 1295 ≥98%
Supplier: Enzo Life Sciences
270-035-M001 270-035-M005
89147-102EA 49.44 USD
89147-102 89147-104
Tyrphostin AG 1295 ≥98%
Tyrphostin AG 1295
Selective inhibitor of tyrosine kinase in platelet-derived growth factor (PDGF) receptor.
CAS Number: 71897-07-9
https://us.vwr.com/stibo/web/std.lang.all/74/59/45347459.jpg