Ebelactone A ≥99%
Supplier: Enzo Life Sciences
270-013-M001
89147-046EA
136.66
USD
89147-046
Ebelactone A ≥99%
Ebelactone A
Esterase inhibitor
Inhibitor of esterases, lipases and N-formylmethionine aminopeptidase.
Microbial.
CAS Number:
76808-16-7 |