Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride) ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: Bestatin hydrochloride(S)-2-((2S,3R)-3-Amino-2-hydroxy-4-phenylbutanamido)-4-methylpentanoic acid hydrochloride

260-012-M005
89146-582EA 110.74 USD
89146-582
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride) ≥98% (by HPLC)
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride)

Inhibits aminopeptidases like aminopeptidase B, leucine aminopeptidase, tripeptide aminopeptidase, aminopeptidases on the surface of mammalian cells. Does not inhibit carboxypeptidases. May be used in in vitro translation system to inhibit the aminopeptidases found in reticulocate or wheat germ lysates. Activates macrophages and T lymphocytes. Immunomodulator. Has antitumor properties and enhances not only blastogenesis and lymphocytes in vitro, but also establishes a delayed-type hypersensitivity in vivo.


  • Aminopeptidase inhibitor


White to off-white powder.


Soluble in methanol, acetic acid, DMSO, dimethyl formamide or 100% ethanol; slightly soluble in water.

Formula: C₁₆H₂₅ClN₂O₄
MW: 308.4 · 36.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00058004
CAS Number: 65391-42-6
https://us.vwr.com/stibo/web/std.lang.all/74/45/45347445.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR