Anti-IP3 Receptor Type 1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-169-R100
89145-272EA 634.53 USD
89145-272
Anti-IP3 Receptor Type 1 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human (weak), mouse, rat and dog IP3R-I. Detects a band of ~240kDa by Western blot.

Type: Primary
Antigen: IP3 RECEPTOR TYPE 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR