Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Glucocorticoid Receptor Beta Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-154-R050
Glucocorticoids are a family of steroids necessary for the regulation of energy metabolism, and the immune and inflammatory responses. These compounds exert their effect through their interaction with the glucocorticoid receptor (GR) and that complex's subsequent association with DNA. All normal mammalian tissues examined to date have been shown to contain GR.The human GR exists in two forms, α and β, which are thought to be the result of alternative splicing of a single gene. Sequence analysis indicates that the α and β forms of human GR are 777 and 742 amino acids long, respectively. They are identical up to residue 727, after which they diverge. After ligand binding, the 94kDa GRα isoform has been observed to translocate from the cytoplasm to the nucleus where it regulates gene expression. In contrast, the 90kDa GRβ isoform does not appear to bind either glucocorticoid agonists or antagonists, and has been localized predominantly in the nucleus independent of hormone treatment in some human cell lines. Studies suggest that human GRβ might function as a dominant negative inhibitor of GRα activity.

For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human GRbeta. Detects a band of ~90kDa by Western blot.

Type: Primary
Antigen: GLUCOCORTICOID RECEPTOR BETA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
50 µL 210-154-R050 89145-248 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol GCRB
Antigen Name Glucocorticoid Receptor Beta
Reactivity Human
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Western Blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Form Liquid. In PBS containing 0.05% sodium azide.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty