Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Global Web is currently experiencing issues, technical teams are working to correct the problem.

Anti-ADRA2A Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
Host: Rabbit

For Immunohistochemistry, Western Blot

Recognizes mouse and rat AR alpha-2a. Detects a band of ~45kDa by Western blot.

Type: Primary
Antigen: ADRA2A
Clonality: Polyclonal
Host: Rabbit
Reactivity: Mouse

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL 210-147-R100 89145-236 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol ADRA2A
Antigen Name Adrenergic Receptor Alpha 2A
Reactivity Mouse
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Western Blot Yes
ImmunoChemistry Yes
Antigen Synonyms adrenergic, alpha-2AAR subtype C10, ADRAR, receptor , alpha-2A-, ADRA2A, ADRA2, ZNF32 , alpha-2A-adrenergic receptor, ADRA2R, ALPHA2AAR, OTTHUMP00000058849
Concentration 0.6mg/ml
Purification Epitope-affinity purified
Form Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty