Anti-FKBP12 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-142-R100
89145-226EA 701.74 USD
89145-226
Anti-FKBP12 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse and rat FKBP12. Detects a band of ~12kDa by Western blot.

Type: Primary
Antigen: FKBP12
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR