Anti-beta-COP Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-127-R100
89145-204EA 612.61 USD
89145-204
Anti-beta-COP Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunocytochemistry, Western Blot

Recognizes human, mouse and rat beta-COP. Detects a band of~110kDa by Western blot.

Type: Primary
Antigen: BETA-COP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR