Anti-C-FOS Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-113-C200
89145-182EA 555.83 USD
89145-182
Anti-C-FOS Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Western Blot

Recognizes mouse c-Fos. Detects a band of ~60kDa by Western blot.

Reconstitute with PBS.

Type: Primary
Antigen: C-FOS
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR