Anti-LEP Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-105-R050
89145-166EA 637.66 USD
89145-166
Anti-LEP Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes human, mouse, rat and sheep leptin.

Type: Primary
Antigen: LEP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR