Anti-TRAF3 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-025-R050
89145-140EA 217.44 USD
89145-140
Anti-TRAF3 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry, Western Blot

Recognizes human TRAF3 and TRAF3b. Detects a band of ~64kDa by Western blot. Does not cross-react with other members of the TRAF family.

Type: Primary
Antigen: TRAF3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR