Anti-NALP1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-018-R050
89145-126EA 233.5 USD
89145-126
Anti-NALP1 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human NLRP1/NALP1. Does not cross-react with mouse NLRP1/NALP1.

Type: Primary
Antigen: NALP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR