Anti-CASP9 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-015-R050
89145-120EA 206.8 USD
89145-120
Anti-CASP9 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat and dog caspase-9. Detects both procaspase-9 and the 35kDa large subunit of activated caspase-9. Detects endogenous protein by IHC and WB.

Type: Primary
Antigen: CASP9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR