Anti-BID Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-006-R050
89145-102EA 191.57 USD
89145-102
Anti-BID Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse and rabbit Bid. Detects a band of ~22kDa by Western blot.

Type: Primary
Antigen: BID
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR