Annexin V (mouse), (recombinant) (Alexa Fluor® 488 conjugate), Enzo Life Sciences
209-257-T100
89145-086EA 356.34 USD
89145-086
Annexin V (mouse), (recombinant) (Alexa Fluor® 488 conjugate), Enzo Life Sciences
Alexa Fluor® 488-labelled recombinant mouse annexin shows bright green fluorescence (Ex(max): 495nm; Em(max): 519nm).
Ordering information: Quantity: 100 tests/500µl.