Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SUPERKILLERTRAIL® (Soluble) (Mouse), (Recombinant), Enzo Life Sciences

Supplier: Enzo Life Sciences
201-130-C020 201-130-3020
Produced in E. coli. The extracellular domain of mouse TRAIL (aa 99-291) is fused at the N-terminus to a His-tag and a linker peptide. The active multimeric conformation is stabilized by an inserted mutation allowing an additional CC-bridge.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
20 µg 201-130-3020 89144-136 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

20 µg 201-130-C020 89144-138 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name SuperKillerTRAIL™ (soluble)
Protein/Peptide Type Recombinant
Species Mouse
Source E. coli
Purity ≥95% (SDS-PAGE)
Endotoxin Content \<10.0EU/mg purified protein (LAL test; Bio Whittaker).
Concentration 0.5mg/ml.
Formulation Liquid. In 20mM HEPES, pH 7.4, 300mM sodium chloride, 0.01% Tween 20, 1% sucrose, 1mM DTT.
Biological Activity Induces apoptosis at concentrations of >10ng/ml as tested on human tumor cells. This effect can be efficiently blocked by recombinant mouse TRAIL receptors (e.g. Prod. No. ALX-522-067 (rmTRAIL-R2:Fc)).Note: This protein does not require a cross-linker (en
Protein Type Recombinant
Purification Affinity purified. Sterile filtered.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty