Human Recombinant KillerTRAIL™ (soluble) (from E. coli)
Supplier: Enzo Life Sciences
201-123-C500
89144-126EA 1970.2 USD
89144-126
Human Recombinant KillerTRAIL™ (soluble) (from E. coli)
Proteins and Peptides
Produced in E. coli. The extracellular domain of human TRAIL (aa 95-281) is fused at the N-terminus to a His-tag and a linker peptide.