Human Recombinant MMP24 (catalytic domain) (from E. coli), HIS Tag
Supplier: Enzo Life Sciences
201-105-C010
89144-104EA 1132.83 USD
89144-104
Human Recombinant MMP24 (catalytic domain) (from E. coli), HIS Tag
Proteins and Peptides
Produced in E. coli.
Activation generates two molecular species. One species begins at the N-terminus with Tyr156 of full-length MMP-24, while the other species is two aa residues shorter and starts with Leu158. Both species extend to Ser351 of MMP-24 and contain an additional His-tag sequence at the C-terminus.