Human Recombinant MMP-14 proenzyme (soluble) (from E. coli), HIS Tag
Supplier: Enzo Life Sciences
201-100-C010
89144-096EA 1229.82 USD
89144-096
Human Recombinant MMP-14 proenzyme (soluble) (from E. coli), HIS Tag
Proteins and Peptides
Produced in E. coli. Prodomain, catalytic domain and hemopexin domain of human MMP-14 proenzyme (aa 1-501) are fused to a His-tag (Thr-(His)6).
Active soluble MMP-14 is used to study the activation of MMP-2 (progelatinase A) and the degradations of proteins of the extracellular matrix, including fibrillar collagens. Proenzyme is activated by trace amounts of MMP-14 catalytic domain.