Human Recombinant SUMO2 (1-95) (from E. coli)
Supplier: Enzo Life Sciences
201-088-C250
89144-064EA 202.83 USD
89144-064
Human Recombinant SUMO2 (1-95) (from E. coli)
Inactive SUMO-2 precursor
Produced in E. coli.
Inactive precursor of human SUMO-2. Substrate for the characterization of SUMO-2 specific proteases.