Human Recombinant Phosphoinositide 3-kinase p110gamma
Supplier: Enzo Life Sciences
201-055-C010
89144-000EA 634.49 USD
89144-000
Human Recombinant Phosphoinositide 3-kinase p110gamma
Enzymes
Produced in Sf9 cells. Full length PI(3)K p110γ (human phosphoinositide-3-kinase p110γ) is fused at the N-terminus to a His-tag.