[pTyr527]pp60c-src (521-533)
Supplier: Enzo Life Sciences
165-022-M001 165-022-MC05
89143-714EA 73.55 USD
89143-714 89143-710
[pTyr527]pp60c-src (521-533)
Proteins and Peptides
This peptide is derived from the C-terminal region of the c-Src protooncogene pp60c-src and binds intramolecularly or intermolecularly to the SH2 domain of the same protein.