Human EGF receptor (652-657)
Supplier: Enzo Life Sciences
155-023-M005 155-023-M001
89143-340EA 75.58 USD
89143-340 89143-342
Human EGF receptor (652-657)
Proteins and Peptides
Specific PKC substrate
Specific substrate (aa 652-657 of mature EGF receptor, without signal peptide (aa 1-24)) for protein kinase C (PKC).