L(+)-Cysteine hydrochloride ≥98% (by TLC)

Synonyms: (R)-2-Amino-3-mercaptopropanoic acid hydrochlorideL-Cysteine hydrochlorideH-Cys-OH.HCl(R)-(+)-Cysteine hydrochloride

101-012-G010
89142-496EA 105.66 USD
89142-496
L(+)-Cysteine hydrochloride ≥98% (by TLC)
L(+)-Cysteine hydrochloride

  • Formulation: White to off-white solid.

Formula: HSCH₂CH(NH₂)COOH·HCl
MW: 157.62 g/mol
MDL Number: MFCD00064553
CAS Number: 52-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR