Anti-HGF Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
905-165 905-165-100
200060-572EA 464.15 USD
200060-572 CA200060-572
Anti-HGF Mouse Monoclonal Antibody
Antibodies
HGF/SF (Hepatocyte growth factor/Scatter factor) is a multifunctional heterodimeric polypeptide. It mediates the growth and scattering of various cell types, epithelial mesenchymal transition, the formation of tubules and lumens, and promotes angiogenesis. The ligand-receptor pair has also shown to be uniquely involved in most human solid tumors and to participate in tumor development, invasion, and metastasis.

  • For Immunohistochemistry, Western Blot

Recognizes human and mouse HGF/SF.

Type: Primary
Antigen: HGF
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR