Glutathione (GSSG/GSH), Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-160
89141-234EA 556.38 USD
89141-234
Glutathione (GSSG/GSH), Detection Kit, Enzo Life Sciences
The Glutathione (Total), detection kit provides a highly sensitive enzymatic recycling reaction for the measurement of total, oxidized, or reduced glutathione in a convenient 96-well plate format.
  • Easy to use sample handling protocols for a wide variety of matrices
  • Convenient and colorimetric 96 well plate format
For the measurement of total glutathione cell lysates, plasma, saliva, tissue, urine, and whole blood from any species.
Delivery information: Supplied with microtiter plate, assay buffer concentrate, reaction mix, glutathione reductase, GSSG.