Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
BSA solution (10%), (50 mL), Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-80-1928 80-1928
95045-196EA 78.46 USD InStock
95045-196 CA95045-196
BSA solution (10%), (50 mL), Enzo Life Sciences
Proteins and Peptides
This BSA Solution (10%) is specially designed to create the Blocking Buffer and Assay Buffer in ImmunoSet products. Each bottle is sufficient for use with 5x96 wells microtiter plates if following the instructions in our ImmunoSet protocol. The BSA Solution may be used for any other assay development needs. Additional assay development reagents are available as companion products to our ImmunoSet products.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty