Sodium hydroxide 5.0 N volumetric solution
Supplier: Hach
|
Danger
|
Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
2450-32
245032
97026-576EA
23.74
USD
CA2450-32
97026-576
Sodium hydroxide 5.0 N volumetric solution
Sodium hydroxide
Multi-purpose sodium hydroxide solution for pH adjustment.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Formula:
NaOH MW: 40 g/mol |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 UN: 1824 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...