Refrigerated Humidity Test Chambers, SHEL LAB
SHC10R SHC28R-2 SHC28R SHC10R-2
89409-508EA 24614.17 USD
89409-508 89409-518 89409-516 89409-510
Refrigerated Humidity Test Chambers, SHEL LAB
Testing Chambers Humidity Chambers
SHEL LAB Refrigerated Humidity Test Chambers provide a controlled environment for a wide range of industrial and biotechnical testing applications
A low pressure water vapor generator, injecting saturated water vapor into the recirculating air duct, controls chamber humidification. This process is preferable to steam generation because steam introduces additional heat to the chamber atmosphere, which then compromises temperature control. Uniform temperature and humidity are maintained by circulating chamber atmosphere through a recirculating air duct system. The air ducts also serve as shelf supports and can be easily removed for cleaning.
Additional standard features include, LED display, microprocessor control, the option of three or six seamless corrosion-resistant stainless steel shelves, and a high limit back-up controller.
This unit incorporates a refrigeration system that dramatically increases the operational range of the cabinet.
Ordering information: Chambers are supplied with manufacturer's two-year limited warranty.