Stablcal® Turbidity Standard, < 0.1 NTU, 500mL, Hach
Supplier: Hach
Stablcal®
2659749 2659749
97026-434EA 68.73 USD InStock
CA11016-414 97026-434
Stablcal® Turbidity Standard, < 0.1 NTU, 500mL, Hach
Standards Turbidity Standards
Stablcal® Turbidity Standard for all Turbidimeters.
  • Precise, Premixed Formazin Solutions
  • Based on Formazin Primary Standards
  • Enhanced Stability Saves Time and Labor
  • Confidence in results
USEPA accepted for calibration purposes. Stablcal® Stabilized Formazin Turbidity Standards are true Formazin dilutions developed for use in any turbidimeter. NTU Value: < 0.1.