Handbook, Microbiology of Well Biofouling, Hach
Supplier: Hach
2617800
89231-152EA 268.66 USD
89231-152
Handbook, Microbiology of Well Biofouling, Hach
Books
A useful reference guide written with a direct, straight forward approach based upon the advances in the technology over the last twenty years.
  • Deleterious events in all types of wells in greater detail
  • Generation of mass which interferes with the physical functioning of a well
  • Major innovations in the field
  • Includes more field applicable material
With this guide, you will understand the phenomena of biofouling, corrosivity, biodegradation, and shifts in hydraulic transmissivity that can be linked to microbial events. Practical approaches to the evaluation of these effects are introduced, including standard and novel methodologies.