Would you like to log in again?
Avantor is taking action. Our response to the Novel Coronavirus outbreak.
Ampule Breaker, Hach
Supplier: Hach
2564000 2564000
97026-758EA 22.48 USD InStock
CA97026-758 97026-758
Ampule Breaker, Hach
Cutters
Breakers help speed testing, and prevent contamination.
Insert ampule into breaker, and press sides together. Ampule tip snaps off.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty