Sodium metaarsenite solution 5.0 g/L

Supplier: Hach

104732 104732
97026-662EA 32.59 USD
97026-662 CA1047-32
Sodium metaarsenite solution 5.0 g/L
Sodium metaarsenite

For sample dechlorination prior to analysis.

Formula: NaAsO₂
MW: 129.91 g/mol
MDL Number: MFCD00003472
CAS Number: 7784-46-5
UN: 2027
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR