Iron and Manganese Color Disc Test Kit, Model IR-20, Hach
Supplier: Hach
146300 1463-00
89231-238EA 246.24 USD InStock
89231-238 CA1463-00
Iron and Manganese Color Disc Test Kit, Model IR-20, Hach
Environmental Test Kits
When customers complain of stained and dingy laundry, this kit lets you do quick, on-the-spot tests for iron and manganese to see where the problem lies.
  • Simple, fast, color disc method
  • Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field
Method/Chemistry: Color disc colorimetry. Test: Iron/ Manganese. Range (mg/L): 0-5/ 0-3. Smallest Increment (mg/L): 0.1/ 0.1. Number of Tests: 100/ 100.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.