Lead Standard Solution, 10 mg/L as Pb (NIST), 25mL, Hach
Supplier: Hach
2374820
89231-060EA 19.36 USD InStock
89231-060
Lead Standard Solution, 10 mg/L as Pb (NIST), 25mL, Hach
Standards Environmental Analysis Standards
For accuracy checks in lead determinations.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.