Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cryogenic Vials, 2.0mL, Color Cap, Corning

Supplier: Corning
431416 431417 431419 431420 431421 431418
30617-556CS 472.34 USD InStock
30617-556 30617-558 30617-562 30617-564 30617-566 30617-560
Cryogenic Vials, 2.0mL, Color Cap, Corning
Vials Cryogenic Vials
These sterile cryogenic vials are manufactured from polypropylene to withstand temperatures to -196 °C.

  • Black graduations and larger marking spots for easier filling and sample identification
  • Color caps
  • Round bottom, self-standing
  • Internally threaded
  • Silicone washer for a secure seal

Certifications: Certified nonpyrogenic and DNase/RNase-free.

Ordering information: 30617-556 comes in assorted colors (Yellow, Blue, Green, Red and White). Each color comes in two bags of 50.

Order Now
 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty