Partisil® PAC HPLC Columns, USP-L18, 5 and 10 μm, Mac Mod
4225-001 4235-001 4230-126
101946-498EA 1644.75 USD
101946-498 101946-510 101946-528
Partisil® PAC HPLC Columns, USP-L18, 5 and 10 μm, Mac Mod
Partisil PAC is a polar amino cyano bonded phase with secondary amine groups for good thermal and chemical stability.
Partisil is a high-purity irregular silica gel available in both 5 and 10 µm particle sizes with a pore size of 80 Å. The choice of column packing includes Silica, C-18 polymeric phases (ODS-3, ODS-2) and C-8. Also available are SAX, SCX, and PAC. These columns provide reproducible results, column to column, lot to lot.
Partisil PAC columns are available only in the fixed endfitting WCS hardware and Magnum-9 hardware.
Selectivity and rapid equilibrium allow a range of separation mechanisms to be used, including adsorption, reversed phase and weak anion exchange. Extremely fast equilibration across the entire range of solvents from heptane to water. The media of choice for carbohydrate separations.
Ordering information: Please see the Partisil/Partisphere accessories page for guard cartridges and hardware kits.